Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse VEGF-B186 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3371
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3371 500 μg
MAB3371SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3371 Supplier R&D Systems Supplier No. MAB3371
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-B Monoclonal specifically detects VEGF-B in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen VEGF-B
Applications Western Blot
Classification Monoclonal
Clone 67409
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49765
Gene Alias vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Gene Symbols VEGFB
Host Species Mouse
Immunogen E. coli-derived recombinant human VEGF-B 186 Accession # P49765
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7423
Test Specificity Detects human VEGF-B 186 in direct ELISAs and Western blots. In these formats, this antibody shows 100% cross-reactivity with recombinant mouse (rm) VEGF-B 186 but does not detect rh or rmVEGF-B 167 rhVEGF 206 rhVEGF 167 rhVEGF-C, rhVEGF-D, rhCTGF, rhPlGF, or rhLDGF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.