Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse WASF1/WAVE1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5514
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5514 Supplier R&D Systems Supplier No. AF5514
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

WASF1/WAVE1 Polyclonal specifically detects WASF1/WAVE1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen WASF1/WAVE1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q92558
Gene Alias KIAA0269WAVEhomology of dictyostelium scar 1, Protein WAVE-1, SCAR1, SCAR1WASP family protein member 1, Verprolin homology domain-containing protein 1, WAS protein family, member 1, WASP1, WAVE1, WAVE1FLJ31482, wisk
Gene Symbols WASF1
Host Species Goat
Immunogen E. coli-derived recombinant human WASF1/WAVE1 Met1-Tyr250 Accession # Q92558
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8936
Test Specificity Detects endogenous human and mouse WASF1/WAVE1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.