Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse WASF3/WAVE3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5515 Supplier R&D Systems Supplier No. AF5515
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

WASF3/WAVE3 Polyclonal specifically detects WASF3/WAVE3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen WASF3/WAVE3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9UPY6
Gene Alias Brush-1, KIAA0900WASP family Verprolin-homologous protein 3, Protein WAVE-3, SCAR3, SCAR3WASP family protein member 3, Verprolin homology domain-containing protein 3, WAS protein family, member 3, WAVE3, WAVE3Brush-1, wisk
Gene Symbols WASF3
Host Species Goat
Immunogen E. coli-derived recombinant human WASF3/WAVE3 Met1-Lys180 Accession # Q9UPY6
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10810
Test Specificity Detects endogenous human/mouse WASF3/WAVE3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.