Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse WDR5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5810
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5810 Supplier R&D Systems Supplier No. AF5810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

WDR5 Polyclonal specifically detects WDR5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen WDR5
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P61964
Gene Alias BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5
Gene Symbols WDR5
Host Species Goat
Immunogen E. coli-derived recombinant human WDR5 Met1-Lys120 Accession # P61964
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11091
Test Specificity Detects human and mouse WDR5 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.