Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Wnt-10a Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB34691SP
This item is not returnable.
View return policy
Description
Wnt-10a Monoclonal specifically detects Wnt-10a in Human, Mouse samples. It is validated for Western Blot.Specifications
Wnt-10a | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P70701 | |
WNT10A | |
E. coli-derived recombinant mouse Wnt-10a Leu55-Glu95 and Gln278-Arg360 Accession # P70701 | |
25 μg | |
Primary | |
Detects mouse and human Wnt-10a in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Wnt-1, -2, -2b, -3a, -4, -5a, -5b, recombinant human (rh) Wnt-6 or rhWnt-7a is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
342811 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ14301, protein Wnt-10a, SSPS, wingless-type MMTV integration site family, member 10A, Wnt10a | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
80326 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction