Learn More
Human/Mouse Wnt-11 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Wnt-11 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Gene Accession No. | Q059Y4 |
| Gene Alias | HWNT11, MGC141946, MGC141948, protein Wnt-11, wingless-type MMTV integration site family, member 11, Wnt11 |
| Gene Symbols | WNT11 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.