Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Wnt-3a Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC13242G100UG
This item is not returnable.
View return policy
Description
Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Wnt-3a | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a | |
E.coli-derived recombinant mouse Wnt-3a Ser36-Gln75, Trp219-Arg269 Accession # P27467 | |
RUO | |
89780 | |
Human, Mouse | |
IgG2a |
Flow Cytometry | |
217804.2R | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P27467 | |
Rat | |
100 μg | |
Primary | |
Detects mouse Wnt-3a in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction