Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Wnt-3a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB13242100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13242100 100 μg
MAB13242SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB13242100 Supplier R&D Systems Supplier No. MAB13242100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Wnt-3a
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 217804.2R
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P27487
Gene Alias MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a
Host Species Rat
Immunogen E.coli-derived recombinant mouse Wnt-3a Ser36-Gln75, Trp219-Arg269 Accession # P27467
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89780
Test Specificity Detects mouse Wnt-3a in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.