Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Mrc2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000149
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000149 Supplier R&D Systems Supplier No. FAB5770G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Mrc2
Applications Flow Cytometry
Classification Monoclonal
Clone 1063704
Conjugate Alexa Fluor 488
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9UBG0
Gene Alias CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Mrc2, Gly31-Ala1414, Accession # Q9UBG0
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9902
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.