Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mrc2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$212.50 - $495.50
Specifications
Antigen | Mrc2 |
---|---|
Clone | 1063704 |
Dilution | 0.25 μg/106 cells, 8 to 25 μg/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5770100
![]() |
R&D Systems
MAB5770100 |
100 μg |
Each for $495.50
|
|
|||||
MAB5770SP
![]() |
R&D Systems
MAB5770SP |
25 μg |
Each for $212.50
|
|
|||||
Description
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Mrc2 | |
0.25 μg/106 cells, 8 to 25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
Mouse myeloma cell line, NS0-derived human Mrc2, Gly31-Ala1414, Accession # Q9UBG0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1063704 | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9UBG0 | |
9902 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title