Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MRGPRF Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8396V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8396V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8396V100 Supplier R&D Systems Supplier No. FAB8396V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRGPRF Monoclonal specifically detects MRGPRF in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MRGPRF
Applications Flow Cytometry
Classification Monoclonal
Clone 916219
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_659452
Gene Alias FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor
Gene Symbols MRGPRF
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MRGPRF Met1-Ser343 Accession # NP_659452
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116535
Test Specificity Detects HEK293 human embryonic kidney cell line transfected with human MRGPRF by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.