Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MRGPRF Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8396R100UG
This item is not returnable.
View return policy
Description
MRGPRF Monoclonal specifically detects MRGPRF in Human samples. It is validated for Flow Cytometry.Specifications
MRGPRF | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_659452 | |
MRGPRF | |
NS0 mouse myeloma cell line transfected with human MRGPRF Met1-Ser343 Accession # NP_659452 | |
RUO | |
116535 | |
Human | |
IgG1 |
Flow Cytometry | |
916219 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor | |
Mouse | |
100 μg | |
Primary | |
Detects HEK293 human embryonic kidney cell line transfected with human MRGPRF by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction