Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MRGX2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4727N100UG
This item is not returnable.
View return policy
Description
MRGX2 Monoclonal specifically detects MRGX2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
MRGX2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q96LB1 | |
MRGPRX2 | |
NS0 mouse myeloma cell line transfected with human MRGX2 Met1-Val330 Accession # Q96LB1 | |
100 μg | |
Primary | |
Detects human MRGX2. Stains human MRGX2 transfectants but not irrelevant transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 °C to 8 °C as supplied. |
Flow Cytometry | |
477533 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGPRX2, MRGX2G protein-coupled receptor MRGX2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
117194 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction