Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MRGX2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4727SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4727SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4727SP Supplier R&D Systems Supplier No. MAB4727SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

MRGX2 Monoclonal specifically detects MRGX2 in Human samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen MRGX2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 477533
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 uL/10^6 cells, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96LB1
Gene Alias MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGPRX2, MRGX2G protein-coupled receptor MRGX2
Gene Symbols MRGPRX2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MRGX2 Met1-Val330 Accession # Q96LB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117194
Test Specificity Detects human MRGX2. Stains human MRGX2 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.