Learn More
Human MS4A3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | MS4A3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 489433 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | NP_006129 |
| Gene Alias | CD20 antigen homolog, CD20 antigen-like protein, CD20L, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTM4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific) |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.