Learn More
Human MS4A4A Alexa Fluor 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | MS4A4A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 818112 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q96JQ5 |
| Gene Alias | 4SPAN1, CD20 antigen-like 1, CD20L1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4, MS4A4MGC22311, MS4A7 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.