Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MS4A4A Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7797S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7797S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7797S100 Supplier R&D Systems Supplier No. FAB7797S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MS4A4A Monoclonal specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MS4A4A
Applications Flow Cytometry
Classification Monoclonal
Clone 818112
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96JQ5
Gene Alias 4SPAN1, CD20 antigen-like 1, CD20L1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4, MS4A4MGC22311, MS4A7
Gene Symbols MS4A4A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0 transfected with human MS4A4A Accession # Q96JQ5
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51338
Test Specificity Detects human MS4A4A.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.