Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MS4A4A PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7797P
This item is not returnable.
View return policy
Description
MS4A4A Monoclonal specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry.Specifications
MS4A4A | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
4SPAN1, CD20 antigen-like 1, CD20L1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4, MS4A4MGC22311, MS4A7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51338 | |
Human | |
Purified |
Flow Cytometry | |
818112 | |
Flow Cytometry 10 uL/10^6 cells | |
Q96JQ5 | |
MS4A4A | |
Mouse myeloma cell line NS0 transfected with human MS4A4A Accession # Q96JQ5 | |
100 Tests | |
Primary | |
Detects human MS4A4A. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction