Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MSP alpha Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB352SP
This item is not returnable.
View return policy
Description
MSP/MST1 Monoclonal specifically detects MSP/MST1 in Human samples. It is validated for Western Blot, Immunoaffinity Purification, ELISA Capture (Matched Antibody Pair).Specifications
MSP/MST1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFL, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, MST1, NF15S2, SF2 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
4485 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Affinity Purification, ELISA | |
45904 | |
Western Blot 1 ug/mL, Immunoaffinity Purification, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
AAA59872 | |
MST1 | |
Mouse myeloma cell line NS0-derived recombinant human MSP Met1-Gly711 Accession # AAA59872 | |
25 μg | |
Primary | |
Detects human MSP alpha Chain in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) HGF or rhIL-6 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction