Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MSP/MST1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF352SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF352SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF352SP Supplier R&D Systems Supplier No. AF352SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

MSP/MST1 Polyclonal specifically detects MSP/MST1 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen MSP/MST1
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.05-0.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA59872
Gene Alias D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFL, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, MST1, NF15S2, SF2
Gene Symbols MST1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MSP/MST1 Gln19-Gly711 Accession # AAA59872
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4485
Test Specificity Detects human MSP/MST1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human HGF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.