Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MSPR/Ron Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF691
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF691 100 μg
AF691SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF691 Supplier R&D Systems Supplier No. AF691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction.

Specifications

Antigen MSPR/Ron
Applications Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL
Gene Accession No. CAA49634
Gene Alias CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4
Gene Symbols MST1R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MSP R Glu25-Ser956 Accession # CAA49634
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4486
Test Specificity Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.