Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MSPR/Ron Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6647F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6647F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6647F Supplier R&D Systems Supplier No. FAB6647F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MSPR/Ron
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. CAA49634
Gene Alias CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4
Gene Symbols MST1R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MSP R/Ron Glu25-Ser956 Accession # CAA49634
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4486
Test Specificity Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.