Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MTF2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6846SP
This item is not returnable.
View return policy
Description
MTF2 Polyclonal specifically detects MTF2 in Human samples. It is validated for Immunohistochemistry.Specifications
MTF2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
M96, metal response element binding transcription factor 2, PCI2, PCL2 | |
Sheep | |
Affinity Purified | |
RUO | |
22823 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
Q9Y483 | |
MTF2 | |
E. coli-derived recombinant human MTF2 Asn356-Ser554 Accession # Q9Y483 | |
25 μg | |
Primary | |
Detects human MTF2 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction