Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MUC-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | MUC1 |
---|---|
Clone | 2336D |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB629812SP
![]() |
R&D Systems
MAB629812SP |
25 μg |
Each for $214.00
|
|
|||||
MAB6298121
![]() |
R&D Systems
MAB629812100 |
100 μg |
Each for $538.00
|
|
|||||
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for ELISASpecifications
MUC1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEM, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
Chinese hamster ovary cell line CHO-derived recombinant human MUC-1, Met1-Ser380, Accession # P15941 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2336D | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
P15941 | |
4582 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title