Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MUC-19 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8245SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8245SP 25 μg
MAB8245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8245SP Supplier R&D Systems Supplier No. MAB8245SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MUC19 Monoclonal specifically detects MUC19 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen MUC-19
Applications Immunohistochemistry
Classification Monoclonal
Clone 876013
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7Z5P9
Gene Alias MUC19, MUC-19
Gene Symbols MUC19
Host Species Mouse
Immunogen E. coli-derived recombinant human MUC-19 Lys5606-Gly5767 Accession # Q7Z5P9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283463
Test Specificity Detects human MUC-19 in direct ELISAs.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.