Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MUC-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8195
This item is not returnable.
View return policy
Description
MUC4 Monoclonal specifically detects MUC4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
MUC-4 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Ascites sialoglycoprotein, ASGP, HSA276359, MUC4, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human MUC4 in ELISA. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
781631 | |
Unconjugated | |
Q99102 | |
MUC4 | |
E. coli-derived recombinant human MUC4 Accession # Q99102 | |
100 μg | |
4585 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction