Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mxi1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4185SP
This item is not returnable.
View return policy
Description
Mxi1 Polyclonal specifically detects Mxi1 in Human samples. It is validated for Immunocytochemistry.Specifications
| Mxi1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHC11, Class C basic helix-loop-helix protein 11, MAD2, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI, MXI-WR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4601 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| P50539 | |
| MXI1 | |
| E. coli-derived recombinant human Mxi1 Ala81-Ser228 Accession # P50539 | |
| 25 μg | |
| Primary | |
| Detects human Mxi1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) MAD1, rhMAD3, and rhMAD4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction