Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Mxi1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4185
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4185 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4185 Supplier R&D Systems Supplier No. AF4185
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Mxi1 Polyclonal specifically detects Mxi1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Mxi1
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. P50539
Gene Alias BHLHC11, Class C basic helix-loop-helix protein 11, MAD2, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI, MXI-WR
Gene Symbols MXI1
Host Species Goat
Immunogen E. coli-derived recombinant human Mxi1 Ala81-Ser228 Accession # P50539
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4601
Test Specificity Detects human Mxi1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) MAD1, rhMAD3, and rhMAD4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.