Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Myeloperoxidase/MPO Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3174
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3174 Supplier R&D Systems Supplier No. AF3174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Myeloperoxidase/MPO Polyclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen Myeloperoxidase/MPO
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1.7-15 ug/mL
Gene Accession No. P05164
Gene Alias EC 1.11.1, EC 1.11.1.7, MPO, myeloperoxidase
Gene Symbols MPO
Host Species Goat
Immunogen Human polymorphonuclear leukocytes Myeloperoxidase and mouse myeloma cell line NS0-derived recombinant human Myeloperoxidase/MPO Ala49-Ser745 Accession # P05164
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Test Specificity Detects human Myeloperoxidase/MPO in direct ELISAs and Western blots. In direct ELISAs, less than 30% cross-reactivity with recombinant mouse MPO is observed and less than 10% cross-reactivity with recombinant human Eosinophil Peroxidase (EPPO) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.