Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MYH6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8979SP
This item is not returnable.
View return policy
Description
MYH6 Monoclonal specifically detects MYH6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
MYH6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-MHC, ASD3, cardiomyopathy, CMD1EE, CMH14, Hypertrophic 1, MYHC, MYHCA, myHC-alpha, Myosin Heavy Chain 6, Myosin Heavy Chain, Cardiac Muscle Alpha Isoform, Myosin, Heavy Chain 6, Cardiac Muscle, Alpha, Myosin, Heavy Polypeptide 6, Cardiac Muscle, Alpha (Cardiomyopathy, Hypertrophic 1), myosin-6, SSS3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4624 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
940344 | |
Western Blot 10 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P13533 | |
MYH6 | |
Human MYH6 peptide Accession # P13533 | |
25 μg | |
Primary | |
Detects human MYH6. The peptide immunogen sequence represents a region specific to MYH6 and not present in other MYH isoforms. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction