Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MYH7 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $503.00

Specifications

Antigen MYH7
Clone 2021A
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 5-25 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB90961SP
GREENER_CHOICE
View Documents
R&D Systems
MAB90961SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB90961100
GREENER_CHOICE
View Documents
R&D Systems
MAB90961100
100 μg
Each for $503.00
Only null left
Add to Cart
 
Description

Description

MYH7 Monoclonal specifically detects MYH7 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

MYH7
Western Blot 0.5 ug/mL, Immunocytochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
cardiac muscle beta isoform, CMD1S, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM
Human MYH7 synthetic peptide Accession # P12883
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
2021A
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
P12883
4625
IgG
Protein A or G purified from cell culture supernatant
Detects human MYH7 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.