Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Myocilin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $538.00
Specifications
Antigen | Myocilin |
---|---|
Clone | 297817 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Myocilin Monoclonal specifically detects Myocilin in Human samples. It is validated for Western Blot.Specifications
Myocilin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q99972 | |
4653 | |
Mouse myeloma cell line NS0-derived recombinant human Myocilin Arg33-Met504 Accession # Q99972 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Myocilin in direct ELISAs and Western blots. In direct ELISAs, 75% cross-reactivity with recombinant mouse Myocilin is observed. |
297817 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GLC1A, GLC1Amyocilin, GPOA, JOAG, JOAG1, MYOC, myocilin, trabecular meshwork inducible glucocorticoid response, TIGR, TIGRmutated trabecular meshwork-induced glucocorticoid response protein, Trabecular meshwork-induced glucocorticoid response protein | |
MYOC | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title