Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Myocilin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB34461100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB34461100 100 μg
MAB34461SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB34461100 Supplier R&D Systems Supplier No. MAB34461100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myocilin Monoclonal antibody specifically detects Myocilin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining)

Specifications

Antigen Myocilin
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 297823
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q99972
Gene Alias GLC1A, GLC1Amyocilin, GPOA, JOAG, JOAG1, MYOC, myocilin, trabecular meshwork inducible glucocorticoid response, TIGR, TIGRmutated trabecular meshwork-induced glucocorticoid response protein, Trabecular meshwork-induced glucocorticoid response protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Myocilin, Arg33-Met504, Accession # Q99972
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4653
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.