Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Myocilin APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB34461A100
This item is not returnable.
View return policy
Description
Myocilin Monoclonal antibody specifically detects Myocilin in Human samples. It is validated for Flow CytometrySpecifications
Myocilin | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
GLC1A, GLC1Amyocilin, GPOA, JOAG, JOAG1, MYOC, myocilin, trabecular meshwork inducible glucocorticoid response, TIGR, TIGRmutated trabecular meshwork-induced glucocorticoid response protein, Trabecular meshwork-induced glucocorticoid response protein | |
Mouse myeloma cell line NS0-derived recombinant human Myocilin, Arg33-Met504, Accession # Q99972 | |
100 Tests | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
297823 | |
Flow Cytometry 10 uL/10^6 cells | |
Q99972 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4653 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction