Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MyoD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | MyoD |
---|---|
Clone | 474515 |
Concentration | LYOPH |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Description
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunocytochemistry.Specifications
MyoD | |
LYOPH | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P15172 | |
4654 | |
E. coli-derived recombinant human MyoD Glu77-Gly215 Accession # P15172 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human MyoD in direct ELISAs. |
474515 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bHLHc1, bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3, MYF3Myf-3, MYOD1, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
MYOD1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title