Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Myoglobin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $491.00
Specifications
Antigen | Myoglobin |
---|---|
Clone | 2269D |
Dilution | Western Blot 0.05 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97204SP
![]() |
R&D Systems
MAB97204SP |
25 μg |
Each for $214.00
|
|
|||||
MAB97204100
![]() |
R&D Systems
MAB97204100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Myoglobin | |
Western Blot 0.05 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
P02144 | |
4151.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2269D | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
MB, MGC13548, myoglobin, PVALB | |
Purified human Myoglobin antigen from human heart, Accession No. P02144 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Myoglobin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title