Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Myoglobin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Myoglobin |
---|---|
Clone | 974145 |
Dilution | Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97202SP
![]() |
R&D Systems
MAB97202SP |
25 μg |
Each for $200.50
|
|
|||||
MAB97202100
![]() |
R&D Systems
MAB97202100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.Specifications
Myoglobin | |
Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
P02144 | |
4151 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
974145 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
MB, MGC13548, myoglobin, PVALB | |
Purified human cardiac myoglobin antigen from human heart Accession # P02144 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Myoglobin in direct ELISAs. In sandwich immunoassays, the antibody pair is specific for human Myoglobin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title