Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Myosin Heavy Chain APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4470A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4470A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4470A Supplier R&D Systems Supplier No. IC4470A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Myosin Heavy Chain
Applications Flow Cytometry
Classification Monoclonal
Clone MF20
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc
Gene Symbols MYH14
Host Species Mouse
Immunogen Chicken pectoralis-derived Myosin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79784
Test Specificity Detects Myosin Heavy Chain in human, mouse, rat and other mammalian, avian, and amphibian species.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.