Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human N-Acetylglucosaminyltransferase V/MGAT5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB5469
This item is not returnable.
View return policy
Description
N-Acetylglucosaminyltransferase V/MGAT5 Monoclonal specifically detects N-Acetylglucosaminyltransferase V/MGAT5 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
N-Acetylglucosaminyltransferase V/MGAT5 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q09328 | |
MGAT5 | |
Mouse myeloma cell line NS0-derived recombinant human N-Acetylglucosaminyltransferase V/MGAT5 Leu189-Leu741 Accession # Q09328 | |
100 μg | |
Primary | |
Detects human N-Acetylglucosaminyltransferase V/MGAT5 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
706824 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A, Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase, EC 2.4.1.155, GGNT5, GlcNAc-T V, GNT-VGNT-VA, Mannoside acetylglucosaminyltransferase 5, mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, MGAT5, N-acetylglucosaminyl-transferase V, N-Acetylglucosaminyltransferase V | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4249 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction