Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human N-Cadherin Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF1388
This item is not returnable.
View return policy
Description
N-Cadherin Polyclonal specifically detects N-Cadherin in Human samples. It is validated for Western Blot.Specifications
N-Cadherin | |
Polyclonal | |
Biotin | |
P19022 | |
CDH2 | |
E. coli-derived recombinant human N-Cadherin Propeptide Ser26-Arg159 Accession # P19022 | |
50 μg | |
Primary | |
Detects human N-Cadherin Propeptide in Western blots. In this format, less than 5% cross-reactivity with recombinant human (rh) N-Cadherin extracellular domain, rhCadherin-8, rhCadherin-11, and rhR-Cadherin is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, NCadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
Sheep | |
Affinity Purified | |
RUO | |
1000 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction