Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human N-Terminal Pro-BNP Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB36042SP
This item is not returnable.
View return policy
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for ELISA.Specifications
BNP | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
E. coli-derived recombinant human Pro-BNP His27-His134 Accession # P16860 | |
25 μg | |
4879 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
ELISA | |
2258B | |
ELISA | |
P16860 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Pro-BNP but does not detect mature human BNP peptide in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction