Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NAALADase-2/NAALAD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7658SP
This item is not returnable.
View return policy
Description
NAALADase-2/NAALAD2 Monoclonal specifically detects NAALADase-2/NAALAD2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NAALADase-2/NAALAD2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9Y3Q0 | |
NAALAD2 | |
Chinese hamster ovary cell line CHO-derived recombinant human NAALADase-2/NAALAD2 Lys32-Leu740 Accession # Q9Y3Q0 | |
25 μg | |
Primary | |
Detects human NAALADase-2/NAALAD2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse NAALADase-2 is observed and no cross-reactivity with recombinant human (rh) PSMA/FOLH1/NAALADase-1, rhNAALADase-like 2, or rhNAALADase-like 1 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
756030 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GCPIII, Glutamate Carboxypeptidase III, GPCIII, MGC116996, MGC26353, NAADALASE2, NAALAD2, NAALADase II, NAALADase2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase II, N-acetylated-alpha-linked acidic dipeptidase 2, N-acetylated-alpha-linked acidic dipeptidase II | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10003 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction