Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NAALADase-2/NAALAD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7658
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
NAALADase-2/NAALAD2 Monoclonal specifically detects NAALADase-2/NAALAD2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NAALADase-2/NAALAD2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9Y3Q0 | |
NAALAD2 | |
Chinese hamster ovary cell line CHO-derived recombinant human NAALADase-2/NAALAD2 Lys32-Leu740 Accession # Q9Y3Q0 | |
100 μg | |
Primary | |
Detects human NAALADase-2/NAALAD2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse NAALADase-2 is observed and no cross-reactivity with recombinant human (rh) PSMA/FOLH1/NAALADase-1, rhNAALADase-like 2, or rhNAALADase-like 1 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
756030 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GCPIII, Glutamate Carboxypeptidase III, GPCIII, MGC116996, MGC26353, NAADALASE2, NAALAD2, NAALADase II, NAALADase2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase II, N-acetylated-alpha-linked acidic dipeptidase 2, N-acetylated-alpha-linked acidic dipeptidase II | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10003 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction