Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NAALADase-like 1/NAALADL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5467
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5467 Supplier R&D Systems Supplier No. AF5467
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NAALADase-like 1/NAALADL1 Polyclonal specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NAALADase-like 1/NAALADL1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. NP_005459
Gene Alias I100, ileal dipeptidylpeptidase, NAALADase L, NAALADase like 1, NAALADASEL, NAALADase-like 1, NAALADL1, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1
Gene Symbols NAALADL1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human NAALADase-like 1/NAALADL1 Ala32-Leu740 Accession # NP_005459
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10004
Test Specificity Detects human NAALADase-like 1/NAALADL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NAALADase I, recombinant mouse NAALADase II, and rhNAALADase II is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.