Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NAALADase-like 2/NAALADL2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB46651SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB46651SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB46651SP Supplier R&D Systems Supplier No. MAB46651SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAALADase-like 2/NAALADL2 Monoclonal specifically detects NAALADase-like 2/NAALADL2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen NAALADase-like 2/NAALADL2
Applications Immunohistochemistry
Classification Monoclonal
Clone 817227
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q58DX5
Gene Alias inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2, NAALADase L2, NAALADase like 2, NAALADL2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Symbols NAALADL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NAALADase-like 2/NAALADL2 Ser152-Asn795 Accession # Q58DX5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254827
Test Specificity Detects human NAALADase-like 2/NAALADL2 in ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.