Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NAIP Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF829
This item is not returnable.
View return policy
Description
NAIP Polyclonal specifically detects NAIP in Human samples. It is validated for Western Blot.Specifications
NAIP | |
Polyclonal | |
Unconjugated | |
NP_004527 | |
NAIP | |
KLH-coupled human NAIP synthetic peptide YLSLSSTRPDEGLASIIC Accession # NP_004527 | |
100 μg | |
Primary | |
Detects human NAIP expressed in Sf 21 insect cells. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
baculoviral IAP repeat-containing 1, BIRC1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP | |
Rabbit | |
Affinity Purified | |
RUO | |
4671 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction