Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NCK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7008
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7008 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7008 Supplier R&D Systems Supplier No. AF7008
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

NCK1 Polyclonal specifically detects NCK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NCK1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P16333
Gene Alias cytoplasmic protein NCK1, MGC12668, NCK adaptor protein 1melanoma NCK protein, nck-1, NCKalpha, NCKNCK tyrosine kinase, non-catalytic region of tyrosine kinase, SH2/SH3 adaptor protein NCK-alpha
Gene Symbols NCK1
Host Species Goat
Immunogen E. coli-derived recombinant human NCK1 Ala2-Ser377 Accession # P16333
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4690
Test Specificity Detects human NCK1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.