Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NCOA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8010
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8010 Supplier R&D Systems Supplier No. AF8010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NCOA2 Polyclonal specifically detects NCOA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen NCOA2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q15596
Gene Alias BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, HTIF2, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2, TIF2hTIF2, Transcriptional intermediary factor 2
Gene Symbols NCOA2
Host Species Sheep
Immunogen E. coli-derived recombinant human NCOA2 Phe1090-Leu1247 Accession # Q15596
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10499
Test Specificity Detects human NCOA2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) NCOA1 is observed, and less than 1% cross-reactivity with rhNCOA4 and rhNCOA6 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.