Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nectin-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2880U100UG
This item is not returnable.
View return policy
Description
Nectin-1/PVRL1 Monoclonal specifically detects Nectin-1/PVRL1 in Human samples. It is validated for Flow Cytometry.Specifications
| Nectin-1/PVRL1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q15223 | |
| PVRL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Nectin-1 Gln31-Thr334 Accession # Q15223 | |
| RUO | |
| 5818 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 610835 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin1, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1, PRR1MGC16207, PVRL1, PVRR1, PVRR1nectin-1, SK-12 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Nectin-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Nectin-2, 3, 4, or recombinant mouse Nectin-1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction