Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nectin-2/CD112 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB2229G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2229G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2229G Supplier R&D Systems Supplier No. FAB2229G
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Nectin-2/CD112
Applications Flow Cytometry
Classification Monoclonal
Clone 610603
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_002847
Gene Alias CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2
Gene Symbols PVRL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112 isoform a Gln32-Leu360 Accession # NP_002847
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5819
Test Specificity Detects human Nectin-2/CD112 by flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.